TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) Biochemistry

Supplier: VWR
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

0694-1L 0694-4L
97062-208EA 125.66 CAD
97062-208 97062-210
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) Biochemistry
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic detergent suited for solubilizing membrane proteins without altering biological activity.

Boiling Pt: 270 °C
Melting Pt: 6
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793

Order Now

Specification Test Results

pH (5%, Water) @25°C 6 - 8
Color (APHA) = 100


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR