Toluidine blue O, high purity
Supplier: VWR
Synonyms:
TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride
0672-25G
0672-50G
97062-166EA
474.13
CAD
97062-166
97062-168
Toluidine blue O, high purity
Toluidine blue O
A blue counterstain often used for metachromatic staining of mast cells, cartilage, and acid mucins.
- Black-gray powder
Formula:
C₁₅H₁₆ClN₃S MW: 305.83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011934 CAS Number: 92-31-9 EINECS: 202-146-2 Merck Index: 12,09658 |
Specification Test Results
Dye Content |
|
Em (626nm, Water) | |
Loss on Drying |
|
Color Index | 52040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...