Sodium sulfite, anhydrous ACS

Supplier: VWR

0628-2.5KG 0628-1KG 0628-500G
97062-058CS 595.19 CAD
97062-058 97062-056 97062-060
Sodium sulfite, anhydrous ACS
Sodium sulphite
Formula: Na₂SO₃
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Chloride 0.02%
Free Acid Test pass
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.005%
Iron = 0.001%
Purity = 98%
Titrable Free Base = 0.03 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR