Coomassie brilliant blue G-250, Ultrapure

Supplier: VWR

Synonyms: Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonate

0615-10G 0615-50G 0615-25G
97062-030EA 97.61 CAD
97062-030 97062-034 97062-032
Coomassie brilliant blue G-250, Ultrapure
Coomassie brilliant blue G-250

Commonly used stain for the detection of protein bands following electrophoresis.

Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1
EINECS: 228-058-4

Order Now

Specification Test Results

Em (595nm, Water) = 36300
Protein Staining Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR