Sodium deoxycholate 99-101%, high purity

Supplier: VWR
Warning

Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate

0613-100G 0613-50G
97062-024CS 3901.51 CAD
97062-024 97062-028
Sodium deoxycholate 99-101%, high purity
Sodium deoxycholate

Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.

Formula: C₂₄H₃₉NaO₄
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7

Order Now

Specification Test Results

Arsenic = 0.001 %
Heavy Metals = 0.005 %
Loss on Drying = 5.0 %
Purity (Dry Basis) 99% - 101 %
Sodium Cholate = 2.0 %
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR