Sodium acetate, anhydrous ACS

Supplier: VWR

Synonyms: Acetic acid sodium salt

0602-2.5KG 0602-1KG 0602-500G
97061-996EA 247.18 CAD
97061-996 97061-994 97061-998
Sodium acetate, anhydrous ACS
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.002%
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.01%
Iron = 0.001%
Loss on Drying = 1%
Magnesium = 0.002%
pH (5%, Water) @25°C 7 - 9.2
Phosphate = 0.001%
Purity = 99%
Sulfate = 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR