Orthoboric acid ACS

Supplier: VWR
Danger

0588-500G 0588-1KG
97061-976CS 585.64 CAD
97061-976 97061-978 97061-974 97061-980
Orthoboric acid ACS
ortho-Boric acid

Biological buffers are useful for cell culture in vitro, enzyme assays, and some electrophoretic applications at physiological pH.

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Heavy Metals (as Pb) = 0.001%
Insolubles (Methanol) = 0.005%
Iron = 0.001%
Nonvolatiles (Methanol) = 0.05%
Phosphate = 0.001%
Purity = 99.5%
Sulfate = 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR