Sodium carbonate, anhydrous ACS

Supplier: VWR
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

0585-2.5KG 0585-500G
97061-970CS 964.15 CAD
97061-970 97061-968 97061-972
Sodium carbonate, anhydrous ACS
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.03%
Chloride = 0.001%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.01%
Iron = 0.0005%
Loss on Heating = 1%
Magnesium = 0.005%
Phosphate = 0.001%
Potassium = 0.005%
Purity = 99.5%
Silica = 0.005%
Sulfur Compounds = 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR