Ethylene glycol, high purity
Supplier: VWR
|
Warning
|
0582-1L
0582-4L
97061-964EA
95.48
CAD
97061-964
97061-966
Ethylene glycol, high purity
Ethylene glycol
Colorless, slightly viscous liquid.Clear with no visible particulates.
Formula:
HOCH₂CH₂OH Boiling Pt: 196…198 °C (1013 hPa) Melting Pt: –13 °C Density: 1.115 g/cm³ (20 °C) Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 13,03832 |
|
Specification Test Results
Acidity |
|
Iron |
|
Moisture (KF) |
|
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...