Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade

Supplier: VWR

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

0571-100KG 0571-500G 0571-2.5KG
97061-934EA 7292.79 CAD
97061-934 97061-942 97061-940
Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: 650
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Heavy Metals (as Pb) 0.001%
Insolubles = 0.03%
Moisture (KF) = 0.3%
pH (1%, Water) @25°C 4.4 - 4.7
Purity = 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR