Diammonium hydrogen orthophosphate ≥98% ACS
Supplier: VWR
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
0569-500G
0569-1KG
0569-2.5KG
97061-932CS
1331.12
CAD
97061-932
97061-928
97061-930
Diammonium hydrogen orthophosphate ≥98% ACS
di-Ammonium hydrogen orthophosphate
Formula:
(NH₄)₂HPO₄ MW: 132.06 g/mol Melting Pt: 185 Density: 1.619 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 EINECS: 231-987-8 REACH: 01-2119490974-22 Merck Index: 13,00546 |
Specification Test Results
Calcium |
|
Chloride |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Magnesium |
|
Nitrate |
|
pH (5%, Water) @25°C | 7.7 - 8.1 |
Potassium |
|
Purity |
|
Sodium |
|
Sulfate |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...