Calcium chloride dihydrate 99-105% ACS

Supplier: VWR
Warning

0556-1KG 0556-2.5KG 0556-500G
97061-900CS 654.33 CAD
97061-900 97061-902 97061-904
Calcium chloride dihydrate 99-105% ACS
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Ammonium 0.005%
Assay 99% - 105%
Barium = 0.005%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.01%
Iron = 0.001%
Magnesium = 0.005%
Oxidizers (as NO3) = 0.003%
pH (5%, Water) @25°C 4.5 - 8.5
Potassium = 0.01%
Sodium = 0.02%
Strontium = 0.1%
Sulfate = 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR