Citric acid, anhydrous ACS

Supplier: VWR
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

0529-3KG 0529-1KG 0529-500G
97061-856CS 1476.24 CAD
97061-856 97061-854 97061-858
Citric acid, anhydrous ACS
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1.665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonizable Substances (H2SO4) Pass
Chloride = 0.001%
Insolubles = 0.005%
Iron = 0.0003%
Lead = 0.0002%
Oxalate Test Pass
Phosphate = 0.001%
Purity = 99.5%
Residue on Ignition = 0.02%
Sulfur Compounds (as SO4) = 0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR