Imidazole ≥99% for biotechnology

Supplier: VWR
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

0527-50G 0527-100G
97064-626EA 94.03 CAD
97064-626 97064-622
Imidazole ≥99% for biotechnology
Imidazole

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
Insolubles = 0.1 %
Melting Point 88 °C - 92 °C
pH (5%, Water) @25°C 9.5 - 10.5
Protease none
Purity 99 %
RNase none detected
Solubility (20%, Water) pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR