PIPES free acid, high purity

Supplier: VWR

Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), Piperazine-1,4-bis(2-ethanesulfonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)diethanesulfonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid, Piperazine-N,N'-bis-2-ethanesulfonic acid

0488-500G 0488-100G
97061-784CS 9420.15 CAD
97061-784 97061-782
PIPES free acid, high purity
PIPES
Formula: C₈H₁₈N₂O₆S₂
Boiling Pt: 499 °C (1013 hPa)
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006159
CAS Number: 5625-37-6
EINECS: 227-057-6
Merck Index: 12,07633
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5192468b680c7bea85a31a131811618356a531b9&vid=ef0e3d488c68ea7f6f583fede6ebf7d2196392b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@260nm (1%, 1N NaOH) ≤ 0.05
DNase none
Heavy Metals (as Pb) 5 ppm
pKa 6.7 - 6.9
Protease none detected
Purity ≥ 98 %
Residue after Ignition ≤ 0.3 %
RNase none detected
Solubility (1%, 1N NaOH) Pass/Fail


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR