Ammonium persulfate (APS) ACS
Supplier: VWR
|
Danger
|
0486-100G
0486-25G
97064-592CS
951.35
CAD
97064-592
97064-594
Ammonium persulfate (APS) ACS
Ammonium peroxodisulphate (APS)
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C Density: 1.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 13,00545 |
|
Specification Test Results
Chloride & Chlorate |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Manganese |
|
Purity |
|
Residue on Ignition |
|
Titrable Free Acid |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...