Sodium thiosulfate pentahydrate, high purity

Supplier: VWR

0484-2.5KG 0484-500G
97061-766CS 846.3 CAD
97061-766 97061-768
Sodium thiosulfate pentahydrate, high purity
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Insolubles 0.005 %
pH (5%, Water) @25 °C 6.0 - 8.4
Purity 99 %
Sulfate & Sulfite 0.1 %
Sulfide 0.0001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR