Coomassie® brilliant blue R-250, Ultrapure
Supplier: VWR
0472-10G
0472-25G
0472-50G
97061-738EA
121.68
CAD
97061-738
97061-740
97061-742
Coomassie® brilliant blue R-250, Ultrapure
Coomassie® Brilliant Blue R-250
Commonly used stain for the detection of protein bands following electrophoresis.
Formula:
C₄₅H₄₄N₃NaO₇S₂ Melting Pt: 174…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041762 CAS Number: 6104-59-2 EINECS: 228-060-5 |
Specification Test Results
Dye Content (Dry Basis) |
|
Em (550nm, Water, Dry Basis) |
|
Loss on Drying | = 5.0 % |
Solubility (0.020g/L, Water) | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...