VWR® Trypsin 1:250, Tissue Culture Grade

Supplier: Avantor
VWR®
VWRV0458-25G VWRV0458-50G VWRV0458-250G
97061-710CS 1740.72 CAD
97061-710 97061-706 97061-708
VWR® Trypsin 1:250, Tissue Culture Grade
Enzymes
Trypsin 1:250, Tissue Culture Grade.

  • EINECS: 232-650-8
  • GHS Symbol 1: GHS08
  • GHS Symbol 2: GHS07
  • MDL: MFCD00082094
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR