Tween® 80 (Polysorbate), Proteomics Grade

Supplier: VWR

0442-1L 0442-4L
97061-674EA 141.11 CAD
97061-674 97061-676
Tween® 80 (Polysorbate), Proteomics Grade
Tween® 80 (Polysorbate)

Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.

Boiling Pt: 100 °C
Melting Pt: –21 °C
Density: 1.062 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 13,07664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR