Cholesterol, Ultrapure

Supplier: VWR

Synonyms: CholesterinCholest-5-en-3-ol(3Beta)-Cholest-5-en-3-ol

VWRV0433-1KG VWRV0433-250G
97061-660CS 3220.01 CAD
97061-660 97061-658
Cholesterol, Ultrapure
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360 °C
Melting Pt: 147 °C…149 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003646
CAS Number: 57-88-5
Merck Index: 13,02221

Order Now

Specification Test Results

Loss on Drying ≤ 0.3 %
Melting Point 146 °C - 150 °C
Residue on Ignition ≤ 0.1 %
Specific Rotation (-38 °)- (-34 °)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR