Lithium chloride for biotechnology

Supplier: VWR
Warning

0416-100G 0416-500G
97061-622CS 884.48 CAD
97061-622 97061-624
Lithium chloride for biotechnology
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Insolubles = 0.01 %
Iron = 10 ppm
Purity 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR