Sodium phosphate dibasic, anhydrous ≥99.0% ACS

Supplier: VWR

Synonyms: di-Sodium hydrogen phosphate anhydrousSodium phosphate dibasicsec-Sodium phosphateDisodium phosphatedi-Sodium hydrogen orthophosphateSodium hydrogen phosphate

VWRV0404-12KG VWRV0404-1KG VWRV0404-2.5KG VWRV0404-500G VWRV0404-50KG
97061-588CS 1268.65 CAD
97061-588 97061-584 97061-586 97061-590 97061-582
Sodium phosphate dibasic, anhydrous ≥99.0% ACS
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00003496
CAS Number: 7558-79-4
Merck Index: 12,08805

Order Now

Specification Test Results

Chloride 0.002%
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.01%
Iron = 0.002%
Loss on Drying @105°C = 0.2%
pH (5%, Water) @25°C 8.7 - 9.3
Purity = 99%
Sulfate = 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR