L(-)-Glutathione (reduced form) ≥98%, high purity

Supplier: VWR

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

0399-250G 0399-50G
97061-572CS 6693.89 CAD
97061-572 97061-574
L(-)-Glutathione (reduced form) ≥98%, high purity
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 192…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

Specification Test Results

Iron = 0.002 %
Lead = 0.001 %
Loss on Drying = 1 %
Purity = 98 %
Specific Rotation -18.5 ° - -15.5 °


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR