Iron(II) sulfate heptahydrate ≥99% ACS

Supplier: VWR

Synonyms: Ferrous sulphate heptahydrateIron vitriolFerrous sulfate heptahydrate

VWRV0387-1KG VWRV0387-500G
97061-542EA 195.68 CAD
97061-542 97061-538
Iron(II) sulfate heptahydrate ≥99% ACS
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64 °C
Density: 1.9 g/cm³ (20 °C)
MDL Number: MFCD00149719
CAS Number: 7782-63-0
Merck Index: 13,04091

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Copper = 0.005%
Ferric Iron = 0.1%
Insolubles = 0.01%
Magnesium = 0.002%
Manganese = 0.05%
Phosphate = 0.001%
Potassium = 0.002%
Purity = 99%
Sodium = 0.02%
Zinc = 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR