Guanidinium thiocyanate ≥99.0% for biotechnology

Supplier: VWR
Danger

Synonyms: Guanidinium sulfocyanideGuanidine ThiocyanateGuanidinium sulphocyanideGTCGuanidinium rhodanide

VWRV0380-500G VWRV0380-250G
97061-522EA 420.9 CAD
97061-522 97061-524
Guanidinium thiocyanate ≥99.0% for biotechnology
Guanidinium thiocyanate

A powerful protein denaturant used to inactivate endogenous RNases during the isolation of RNA from tissues and bacterial cultures

Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 115…118 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00013027
CAS Number: 593-84-0
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Abs.@280nm (70%, Water) = 0.8
Abs.@300nm (6M, Water) 0.1
Abs.@410nm (6M, Water) 0.1
Copper 0.0001 %
DNase none detected
Solubility (70%, Water 50° C) pass
Lead 0.0001 %
Melting Point 118 °C - 121 °C
Protease none detected
Purity 99 %
RNase none detected

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR