CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥99%, high purity

Supplier: VWR

0365-500G 0365-1KG 0365-250G
97061-492CS 1471.12 CAD
97061-492 97061-488 97061-490
CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥99%, high purity
CAPS (3-(Cyclohexylamino)propanesulphonic acid)

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological

Formula: C₆H₁₁NH(CH₂)₃SO₃H
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Order Now

Specification Test Results

pKa @25°C 10.3 - 10.5
Purity = 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR