Sodium pyruvate ≥99%, Reagent Grade
Supplier: VWR
0342-100G
0342-1KG
0342-250G
97061-448CS
2022.49
CAD
97061-448
97061-450
97061-452
Sodium pyruvate ≥99%, Reagent Grade
Sodium pyruvate
Formula:
CH₃COCOONa Melting Pt: 220…230 °C Density: 1.718 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002586 CAS Number: 113-24-6 EINECS: 204-024-4 |
Specification Test Results
Free Acid |
|
Heavy Metals (as Pb) |
|
Water Content |
|
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...