D-(+)-Biotin, Reagent Grade
Supplier: VWR
0340-1G
0340-5G
97061-444EA
126.42
CAD
97061-444
97061-446
D-(+)-Biotin, Reagent Grade
D(+)-Biotin
A small biomolecule, 244 Daltons, that has high binding affinity to avidin and streptavidin often used to label antibodies and enzymes. Several molecules of biotin can react with a single protein molecule to increase the sensitivity at the site of detection.
Formula:
C₁₀H₁₆N₂O₃S Melting Pt: 232 °C Density: 1.41 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005541 CAS Number: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Specification Test Results
Heavy Metals (as Pb) |
|
Purity (RG) |
|
Specific Rotation | 89°- 93° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...