D-(+)-Sucrose, Ultrapure DNase-, RNase-free

Supplier: VWR

0335-12KG 0335-50KG 0335-5KG 0335-500G 0335-2.5KG 0335-10KG 0335-1KG
97061-426EA 955.3 CAD
97061-426 97061-434 97061-436 97061-432 97061-430 97061-424 97061-428
D-(+)-Sucrose, Ultrapure DNase-, RNase-free
D(+)-Sucrose

DNase- and RNase-free.Common reagent in routine nucleic acid procedures and electrophoresis loading dyes.

Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.005%
DNase none detected
Glucose = 0.1%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Invert Sugar = 0.1%
Iron = 0.0005%
Loss on Drying = 0.03%
Protease none detected
Purity = 99.9%
RNase none detected
Solubility (10%, Water) Pass
Specific Rotation 66.3° - 66.8 °
Sulfate = 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR