Bovine Serum Albumin (BSA)
Supplier: VWR
0332-500G
0332-25G
97061-422CS
6426.07
CAD
97061-422
97061-418
97061-420
Bovine Serum Albumin (BSA)
Proteins and Peptides
Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.
- Nuclease-free
- Protease-free
- Heavy Metals (as Pb) <0.001 %
- Iron <0.0005 %
- Purity >98 %
pH (5%, Water) @25 °C 6.5 - 7.5.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...