Nitro blue tetrazolium chloride (NBT), Ultrapure
Supplier: VWR
|
Warning
|
0329-500MG
0329-1G
97061-414EA
222.05
CAD
97061-414
97061-412
Nitro blue tetrazolium chloride (NBT), Ultrapure
Nitro blue tetrazolium chloride (NBT)
Formula:
C₄₀H₃₀Cl₂N₁₀O₆ Melting Pt: 200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012159 CAS Number: 298-83-9 EINECS: 206-067-4 |
Specification Test Results
Em (Lambda Max, Methanol) | > 54000 |
Lambda Max (Methanol) | 255 - 260 nm |
Loss on Drying | < 10.0 % |
Solubility (1%, Water) | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...