EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure
Supplier: VWR
|
Warning
|
0322-1KG
0322-500G
97061-404CS
496.42
CAD
97061-404
97061-406
EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure
EDTA (Ethylenediamine tetraacetic acid)
Calcium chelator used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals.
Formula:
C₁₀H₁₆N₂O₈ Boiling Pt: 614 °C (1013 hPa) Melting Pt: 240 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
|
Specification Test Results
DNase | none detected |
Heavy Metals (as Pb) |
|
Insolubles (5%, 1N NaOH) | Pass |
Iron |
|
Magnesium |
|
Nitrilotriacetic Acid |
|
pH (5%, Water) @25°C | 2.5 - 3.5 |
Protease | none detected |
Purity |
|
Residue after Ignition |
|
RNase | none detected |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...