Formamide, high purity
Supplier: VWR
|
Danger
|
0314-950ML
0314-500ML
97061-394EA
171.59
CAD
97061-394
97061-392
Formamide, high purity
Formamide
Ideal for denaturing nucleic acids in hybridization and DNA sequencing procedures.
Formula:
HCONH₂ Boiling Pt: 210… 216 °C Melting Pt: 1…3 °C Density: 1.1334 g/cm³ (20 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 Merck Index: 13,04261 |
|
Specification Test Results
Abs.@280nm |
|
Conductivity |
|
Copper |
|
Freezing Range | 1 °C - 3 °C |
Iron |
|
Lead |
|
Purity |
|
Zinc |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...