Magnesium chloride hexahydrate ACS

Supplier: VWR

0288-100G 0288-1KG 0288-500G
97061-352CS 808.44 CAD
97061-352 97061-354 97061-356
Magnesium chloride hexahydrate ACS
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Ammonium 0.002%
Barium = 0.005%
Calcium = 0.01%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iron = 0.0005%
Manganese = 0.0005%
Nitrate = 0.001%
Phosphate = 0.0005%
Potassium = 0.005%
Purity 99% - 102%
Sodium = 0.005%
Strontium = 0.005%
Sulfate = 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR