Guanidinium hydrochloride for biotechnology
Supplier: VWR
Synonyms:
Guanidinium chloride
, Guanidine hydrochloride
, Guanidine chloride
, Aminoformamidine hydrochloride
, Guanadine hydrochloride
, Aminomethanamidine hydrochloride
, Guanidine HCI
VWRV0287-500G
VWRV0287-100G
97061-348EA
87.04
CAD
97061-348
97061-350
Guanidinium hydrochloride for biotechnology
Guanidine hydrochloride
Suited for purification of nucleic acids or polypeptides from cell sources and inhibition of both RNase and protease activity.
Formula:
NH₂C(=NH)NH₂·HCl MW: 95.53 g/mol Melting Pt: 180…185 °C Density: 1.354 g/cm³ (20 °C) |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 Merck Index: 13,04578 |
Specification Test Results
Solubility (6M, Water) | pass |
Abs.@260nm (6M, Water) |
|
Arsenic |
|
DNase | none detected |
Iron |
|
Lead |
|
Melting Point | 181 °C - 187 °C |
Protease | none detected |
Purity |
|
RNase | none |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...