Dithiothreitol (DTT, Cleland's reagent) for biotechnology

Supplier: VWR
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

0281-25G 0281-5G
97061-338CS 8369.04 CAD
97061-338 97061-340
Dithiothreitol (DTT, Cleland's reagent) for biotechnology
Dithiothreitol (DTT, Cleland's reagent)

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42 C…43 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@283nm (0.02M, Water) = 0.05
DNase none detected
Loss on Drying 0.5 %
Melting Point 39 °C - 43 °C
Oxidized DTT 0.5 %
Protease none detected
Purity (-SH Content) 99.4 %
RNase none detected
Solubility (5%, Water) pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR