EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade

Supplier: VWR
Danger

Synonyms: Tetrasodium ethylenediaminetetraacetate hydrate, Kestranal® 4S hydrate, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate, Edetic acid tetrasodium salt hydrate, Ethylenediaminetetraacetic acid tetrasodium salt hydrate

0245-500G 0245-1KG 0245-2.5KG
97061-294CS 1014.08 CAD
97061-294 97061-290 97061-292
EDTA tetrasodium salt hydrate ≥99.5%, Ultra Pure Grade
EDTA tetrasodium salt hydrate
Formula: C₁₀H₁₂N₂O₈Na₄·xH₂O
MW: 380.17 g/mol (anhy)
Melting Pt: > 300 °C
Density: 6.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150025
CAS Number: 13235-36-4
EINECS: 200-573-9
Merck Index: 12,03557

Order Now

Specification Test Results

Heavy Metals (as Pb) = 0.001 %
Solubility (5%, Water) pass
Iron = 0.005 %
Moisture (KF) 12.4 % - 19.2 %
Nitrilotriacetic Acid = 0.1 %
pH (1%, Water) @25°C 10.7 - 11.7
Purity (Dry Basis) 99.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR