Sodium chloride for biotechnology

Supplier: VWR

Synonyms: Saline, NaCl, Sea salt, Halite

0241-10KG 0241-1KG 0241-12KG 0241-5KG 0241-2.5KG 0241-500G 0241-50KG
97061-266EA 408.98 CAD
97061-266 97061-270 97061-268 97061-278 97061-264 97061-272 97061-274 97061-276
Sodium chloride for biotechnology
Sodium chloride

Common source of salts for media propagation of many organisms.

Formula: NaCl
Boiling Pt: 1413 °C
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Calcium = 0.005%
DNase none detected
Heavy Metals = 0.0005%
Iodide = 0.002%
Iron = 0.0002%
Loss on Drying = 0.5%
Magnesium = 0.005%
Nitrogen Compounds = 0.001%
pH (5%, Water) @25°C 5 - 9
Phosphate = 0.0005%
Potassium = 0.005%
Protease none detected
Purity = 99.9%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR