Calcium carbonate, anhydrous, Reagent Grade

Supplier: VWR

Synonyms: Marble

VWRV0179-500G VWRV0179-1KG
97061-148CS 705.26 CAD
97061-148 97061-150
Calcium carbonate, anhydrous, Reagent Grade
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825 °C
Density: 2.71 g/cm³ (20 °C)
MDL Number: MFCD00010906
CAS Number: 471-34-1
Merck Index: 13,01658

Order Now

Specification Test Results

Arsenic <= 0.0003 %
Fluoride <= 0.005 %
Heavy Metals <= 0.003 %
Lead <= 0.001 %
Loss on Drying <= 2.0 %
Magnesium and Alkali Salts <= 1.0 %
Purity > 98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR