L(+)-Ascorbic acid free acid ACS

Supplier: VWR

Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

0143-100G 0143-500G
97061-072CS 694.95 CAD
97061-072 97061-074
L(+)-Ascorbic acid free acid ACS
L(+)-Ascorbic acid
Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
Melting Pt: 190…192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals (as Pb) 0.002%
Iron = 0.001%
Purity = 99
Residue on Ignition = 0.1%
Specific Rotation 20.5° - 21.5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR