Guanidinium hydrochloride, Technical Grade
Supplier: VWR
|
Warning
|
0118-500G
0118-1KG
97061-044CS
983.23
CAD
97061-044
97061-042
Guanidinium hydrochloride, Technical Grade
Guanidine hydrochloride
Suited for purification of nucleic acids or polypeptides from cell sources and inhibition of both RNase and protease activity.
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 180…185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Specification Test Results
Ammonium Chloride |
|
Ash |
|
Moisture (KF) |
|
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...