EDTA disodium salt dihydrate for biotechnology

Supplier: VWR
Warning

0105-12KG 0105-1KG 0105-2.5KG 0105-500G
97061-016EA 2275.07 CAD
97061-016 97061-018 97061-020 97061-022
EDTA disodium salt dihydrate for biotechnology
EDTA disodium salt dihydrate

A reversible metalloprotease inhibitor useful for chelating metal ions.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
Heavy Metals (as Pb) = 0.005%
Insolubles = 0.005%
Iron = 0.01%
Loss on Drying 8.7% - 11.4%
Nitrilotriacetic Acid = 0.1%
pH (5%, Water) @25°C 4.0 - 6.0
Protease none detected
Purity = 99%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR