tri-Sodium citrate dihydrate, Reagent Grade

Supplier: VWR

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

0101-2.5KG 0101-1KG 0101-500G 0101-12KG
97061-006CS 487.21 CAD
97061-006 97061-004 97061-008 97061-002
tri-Sodium citrate dihydrate, Reagent Grade
tri-Sodium citrate dihydrate

A biological buffer used for cell culture, in vitro, enzyme assays and some electrophoretic applications at physiological pH.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium = 0.005%
Chloride = 0.005%
DNase none detected
Heavy Metals (as Pb) =0.001%
Insolubles =0.005%
Iron =0.0005%
Loss on Drying (%) 10.0 - 13.0
pH (5%, Water) @25°C 7.0 - 9.0
Protease none detected
Purity (Dry Basis) (%) = 99%
RNase none detected
Sulfate =0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR