CellSmart Tissue Culture Plates, Argos Technologies
Supplier: Antyila Scientific
01927-70 01927-72 01927-74 01927-78 01927-76
89126-806CS 503.76 CAD
89126-806 89126-808 89126-810 89126-812 89126-814
CellSmart Tissue Culture Plates, Argos Technologies
Microplates Cell Culture Plates
Argos cell and tissue culture plates are optically clear for easy visualization of cells
Products are manufactured in a class 100,000 clean room environments under ISO 9001:9002 quality systems. All products are manufactured with 100% USP VI crystal clear virgin polystyrene and high quality PE to ensure the integrity of your cells. Our cell and tissue culture products are specially treated with patented vacuum-gas plasma machine that causes the extremely hydrophobic polystyrene surface to become negatively charged and hydrophilic. This allows the cells to attach more evenly, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable