Disposable Mini Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
22999-30 22999-31 22999-32
89126-860CS 367.56 CAD
89126-860 89126-862 89126-864
Disposable Mini Plastic Serological Pipettes, Argos Technologies
Pipettes Serological Pipettes
Argos brand mini pipettes were developed for use in confined spaces such as laminar hoods
Fiber free paper & plastic wrap ensures quick peel apart opening. Polystyrene, gamma sterilized & pyrogen free. Dispenser box allows for access of pipettes without opening top; color coded with universal stacking design.
These mini pipettes measure 9" and fit most serological pipettors. Smaller size reduces distance between sample and pipettor aiding in more ergonomic pipetting. Unique filter plug stays put and eliminates lint fibers. Pipettes feature color coded rings for ease in identification, easy-to-read graduations from +1 to 2%.