FilterMax™ F3 and F5 Multimode Microplate Readers, Molecular Devices
Supplier: Molecular Devices
FilterMax™
F5 F3 F3-SC F3 F5-SC F5
CA97059-524EA 79881.26 CAD
CA97059-524 97059-524 97059-526 10067-742 CA97059-526 10067-740
FilterMax™ F3 and F5 Multimode Microplate Readers, Molecular Devices
Microplate Readers
The FilterMax™ F3 features absorbance (visible), fluorescence intensity - FI (top reading), and glow luminescence for 96- to 384-well plates.
  • Multiple programmable options
  • Six-position filter slides
  • Linear and orbital shaking
  • Temperature control
Users can program endpoint, kinetic, multiple wavelength, linear scan, and area scan measurement types. Easily accessible six-position filter slides enable wavelength selection from 340 to 650nm.
The FilterMax™ F5 features absorbance (UV and visible), fluorescence intensity - FI (top and bottom reading), glow luminescence, fluorescence polarization (FP), and time-resolved fluorescence (TRF) for 6- to 1536-well plates. In addition, the instrument features linear and orbital shaking, plus temperature control. Users can program endpoint, kinetic, multiple wavelength, linear scan, and area scan measurement types. Easily accessible six-position filter slides enable wavelength selection from 230 to 700nm.
The FilterMax™ F-Series of readers can operate as standalone instruments or they can be integrated seamlessly with laboratory automation platforms. Units are ideal for a broad range of applications, including drug discovery, genomics, proteomics, and cell-based research. The unique patented design ensures precise performance and sensitivity across all detection modes. Units can use either automated or manual Z-Height Optimization, and can utlize high-density well scanning for unique applications.
The FilterMax™ F-Series of readers are the first microplate readers to employ patented high-power Light Emitting Diodes (LED). The FilterMax™ is able to optimize the signal at the light source for specific dyes or applications, providing high power at critical wavelengths. The Fluorescence Detector Photomultiplier (PMT) operates in photon counting mode, affording increased sensitivity for red dyes and a linear dynamic range over 6 orders of magnitude.
Ordering information: Instrument includes Excitation Filter Slide #3 (Absorbance: 405nm, 450nm, 492nm, 620nm. Fluorescence: 360nm, 485nm), Emission Filter Slide #3 (Fluorescence: 465nm, 535nm), current version of SoftMax Pro Software, power cable, RS-232 serial cable, and user manuals. Standard 12-month warranty included. Purchase of additional One Year Service Plan includes MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling, along with unlimited technical support and loaner instrument (if available), are provided with this service option.