AquaMax® Microplate Washers, Molecular Devices
Supplier: Molecular Devices
AQUAMAX 2K BUNDLE AQUAMAX 4K BUNDLE 0310-5363 0310-5227
97059-514EA 34829.61 CAD
97059-514 97059-518 CA89429-526 CA89429-528
AquaMax® Microplate Washers, Molecular Devices
Microplate Washers
AquaMax® Microplate Washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
  • Interchangeable heads
  • One-Tough AutoClean feature
  • Includes 1 year parts and labor manufacturer's warranty
Wash heads can be changed in 30 seconds with no tools using the innovative head latch mechanism
AquaMax® Washers have several important maintenance features, since the lack of proper washer maintenance is the number one cause of washer failure. Using the One-Touch AutoCleanMore Product Information