uTIP™ Pipette Tips for Universal Pipettors, Biotix
Supplier: Biotix
uTIP™
M-0200-9NC 63305452 M-0010-9NC M-0200-1RCNS M-0250-9NC M-1000-BC M-1250-9SC96 M-0011-9NC M-0011-9SC M-1000-9SC M-0300-9NC M-1000-9NC M-0250-BC M-0200-9SC M-1250-9NC M-0250-9SC M-0010-9SC M-0300-9SC 63305454 63305451 M-0200-1RC 63305453
76175-408CS 409.42 CAD
76175-408 76175-414 77575-930 10027-736 76175-388 76175-410 10027-668 89425-644 76175-382 89425-646 76175-282 10027-672 10027-670 10027-666 89408-548 89136-570 77575-924 89136-574 77575-926 89136-566 77575-928 89136-578
uTIP™ Pipette Tips for Universal Pipettors, Biotix
Pipette Tips
Biotix® uTIP Racked- Non-Filtered, Low Retention, Pre-Sterilized Universal pipette Tips are designed to improve ergonomics by lowering the necessary insertion and ejection force by 66 to 80% to make a secure seal. This is achieved through the patented Flexfit™ and Blade™ technologies. All Biotix® tips are manufactured with X-Resin™, a proprietary ultra-low retention resin which results in tighter CV values.
  • The FlexFit™ seal is able to be secured with just the weight of a pipettor
  • Blade™ Technology lowers the chance of hanging droplet formation by optimizing surface area at the distal end of the tip
  • Blade™ Technology eliminates the need for tip touch off, saving time with each draw
Biotix® uTIP™ pipette tips fit all standard pipettors and are designed to improve ergonomics by lowering insertion and ejection forces by 66 to 80%.
All Biotix® tips are certified free of RNase, DNase, Pyrogens, and PCR Inhibitors. They are manufactured under stringent controls in an ISO 9001 certified facility. Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected for quality. All racks include a label with a unique bar-coded serial number, part number, and description for traceability to the individual rack.
X-Resin®, a proprietary ultra-low retention resin, ensures minimal sample retention in the tip, improving the accuracy and precision of all results.
Chem-Resin™ provides a solution for working with challenging strong polar solvents or highly electronegative reagents. The tips have a more neutral surface tension. This reduced hydrophobicity on the surface facilitates low volume draw and drip-free dispense of these classes of chemicals.