DNA Markers and Ladders
Supplier: Avantor
50321 50323 50461 50471 50322 50330 50327 50333
CA12001-496EA 210.05 CAD
CA12001-496 CA12001-498 CA12001-500 CA12001-502 CA12001-504 CA12001-730 CA12001-732 CA12001-738 CA12001-776 CA12001-778 CA12001-780 CA12001-782 CA12001-784 CA12001-786 CA12001-810
DNA Markers and Ladders
Electrophoresis Reagents Molecular Weight Markers
Lonza DNA Ladders are precision manufactured to provide rapid and easy size determination of your DNA samples.
  • Ideal for PCR or cloning verification
  • Plasmid-free
Lonza SimplyLoad™ DNA ladders are ready-to-load directly on your gel to maximize your time.
Lonza’s standard DNA ladders are ready-to-dilute prior to loading on your gel. Plasmid-free to ensure minimal background.
For all of Lonza’s ladders, each band has an ideal 50% G-C content for rapid, precise sizing and sharp resolution. Our ladders are free from contaminating plasmid DNA so they have minimal background, giving you the perfect gel the first time, every time.
SimplyLoad™ DNA ladders are supplied at ready-to-load concentrations in TE Buffer and 1X Ficoll® sample loading buffer containing Bromophenol blue and Xylene cyanol FF so you can get straight to your results.